Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYOG SIRNA Assays ›
          Gene Symbol:
          Myog
          Gene Name:
          myogenin
          Gene Aliases:
          MYF4, bHLHc3, myo
          Chromosome Location:
          Chr.1: 134290004 - 134292548 on Build GRCm38
          UniGene ID:
          Mm.16528
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 84974

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Myog
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031189.2 NP_112466.1 1 338
          GenBank mRNA AB221643.1 - 1 286
          BC048683.1 - 1 353 6516364
          BC068019.1 - 1 319 6508229
          CJ096370.1 - 1 338
          D90156.1 - 1 329
          X15784.1 - 1 334

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017115, AF054894, M24393.

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cell cycle
          multicellular organism development
          muscle organ development
          skeletal muscle tissue development
          negative regulation of cell proliferation
          positive regulation of myotube differentiation
          positive regulation of muscle atrophy
          regulation of skeletal muscle satellite cell proliferation
          response to muscle activity involved in regulation of muscle adaptation
          response to electrical stimulus involved in regulation of muscle adaptation
          striated muscle atrophy
          response to denervation involved in regulation of muscle adaptation
          myotube differentiation
          cell differentiation
          skeletal muscle cell differentiation
          mRNA transcription from RNA polymerase II promoter
          histone H3 acetylation
          histone H4 acetylation
          positive regulation of myoblast differentiation
          negative regulation of glycolytic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          skeletal muscle fiber development
          positive regulation of skeletal muscle fiber development
          positive regulation of cell cycle arrest
          cellular response to lithium ion
          cellular response to growth factor stimulus
          cellular response to estradiol stimulus
          regulation of myoblast fusion
          positive regulation of myoblast fusion
          positive regulation of oxidative phosphorylation
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          core promoter proximal region sequence-specific DNA binding
          core promoter binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          chromatin DNA binding
          sequence-specific DNA binding
          protein dimerization activity
          E-box binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7xtrm:80/100.66.78.85:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0