Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BMPR1B SIRNA Assays ›
          Gene Symbol:
          BMPR1B
          Gene Name:
          bone morphogenetic protein receptor type 1B
          Gene Aliases:
          ALK-6, ALK6, AMDD, BDA1D, BDA2, CDw293
          Chromosome Location:
          Chr.4: 94757977 - 95158453 on Build GRCh38
          UniGene ID:
          Hs.598475
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 85

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 6,301.00
          Your Price
          Online offer:
          6301.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BMPR1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001203.2 NP_001194.1 5 508
          NM_001256792.1 NP_001243721.1 3 404
          NM_001256793.1 NP_001243722.1 3 345
          NM_001256794.1 NP_001243723.1 3 388
          XM_011532201.2 XP_011530503.1 4 387
          XM_017008558.1 XP_016864047.1 5 579
          XM_017008559.1 XP_016864048.1 6 569
          XM_017008560.1 XP_016864049.1 3 461
          XM_017008561.1 XP_016864050.1 4 418
          GenBank mRNA AK293008.1 - 3 388
          AK299930.1 - 3 345
          AK313642.1 - 3 404
          BC047773.1 - 2 304 6202951
          BC069796.1 - 4 420 7262484
          BC069803.1 - 4 420 7262496
          D89675.1 - 2 252
          U89326.1 - 5 507

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          cartilage condensation
          ovarian cumulus expansion
          eye development
          chondrocyte development
          protein phosphorylation
          inflammatory response
          dorsal/ventral pattern formation
          signal transduction by protein phosphorylation
          proteoglycan biosynthetic process
          positive regulation of bone mineralization
          BMP signaling pathway
          retinal ganglion cell axon guidance
          positive regulation of chondrocyte differentiation
          limb morphogenesis
          ovulation cycle
          positive regulation of cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          retina development in camera-type eye
          endochondral bone morphogenesis
          positive regulation of cartilage development
          cellular response to BMP stimulus
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of chondrocyte proliferation
          glycoprotein binding
          protein serine/threonine kinase activity
          transmembrane receptor protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          transforming growth factor beta receptor activity, type I
          protein binding
          ATP binding
          SMAD binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gt9tc:80/100.66.77.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0