Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wdr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031392.2 | NP_113569.1 | 2 | 355 | ||
GenBank mRNA | AB041854.1 | - | 2 | 424 | ||
AF348591.1 | - | 2 | 351 | |||
AK037181.1 | - | 2 | 332 | |||
AK046119.1 | - | 2 | 322 | |||
AK052235.1 | - | 2 | 294 | |||
AK146617.1 | - | 2 | 312 | |||
AK163752.1 | - | 2 | 288 | |||
AK165935.1 | - | 2 | 319 | |||
AK166265.1 | - | 2 | 332 | |||
AK169357.1 | - | 2 | 356 | |||
BC050894.1 | - | 2 | 355 | 5708880 | ||
BC054367.1 | - | 2 | 283 | 5346943 |