Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HSPA8 SIRNA Assays ›
Gene Symbol:
Hspa8
Gene Name:
heat shock protein 8
Gene Aliases:
2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
Chromosome Location:
Chr.9: 40801273 - 40805199 on Build GRCm38
UniGene ID:
Mm.290774
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
85046
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hspa8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031165.4
NP_112442.2
5
667
GenBank mRNA
AK004608.1
-
4
956
AK035286.1
-
5
665
AK075935.1
-
5
664
AK081701.1
-
5
666
AK098074.1
-
5
664
AK145416.1
-
5
587
AK145579.1
-
5
661
AK146547.1
-
5
668
AK146708.1
-
5
662
AK146985.1
-
5
665
AK147864.1
-
5
665
AK149954.1
-
5
662
AK150474.1
-
5
659
AK150498.1
-
5
659
AK150701.1
-
5
669
AK150958.1
-
5
662
AK151041.1
-
5
665
AK151065.1
-
5
665
AK151127.1
-
5
668
AK151287.1
-
5
665
AK151435.1
-
5
665
AK151516.1
-
5
671
AK151537.1
-
5
662
AK151741.1
-
5
665
AK151775.1
-
5
667
AK151808.1
-
5
668
AK151865.1
-
5
665
AK151892.1
-
5
667
AK151948.1
-
5
668
AK151997.1
-
5
665
AK152315.1
-
5
668
AK152598.1
-
5
665
AK152697.1
-
5
665
AK152703.1
-
5
665
AK152768.1
-
5
668
AK152803.1
-
5
668
AK153032.1
-
5
667
AK153834.1
-
5
665
AK157831.1
-
5
665
AK159479.1
-
5
665
AK162775.1
-
5
665
AK163987.1
-
5
665
AK164000.1
-
5
665
AK166545.1
-
5
667
AK166643.1
-
5
679
AK166721.1
-
5
662
AK166767.1
-
5
665
AK166776.1
-
5
668
AK166808.1
-
5
665
AK166830.1
-
5
663
AK166846.1
-
5
665
AK166861.1
-
5
668
AK166873.1
-
5
665
AK166908.1
-
5
668
AK166910.1
-
5
665
AK166913.1
-
5
665
AK166933.1
-
5
665
AK167043.1
-
5
667
AK167121.1
-
5
668
AK167122.1
-
5
665
AK167134.1
-
5
663
AK167163.1
-
5
667
AK167218.1
-
5
665
AK167229.1
-
5
666
AK167254.1
-
5
666
AK167414.1
-
5
665
AK167845.1
-
5
668
AK167910.1
-
5
668
AK168435.1
-
5
668
AK168492.1
-
5
667
AK168511.1
-
3
203
AK168519.1
-
5
665
AK168542.1
-
5
667
AK168577.1
-
5
665
AK168625.1
-
5
665
AK168654.1
-
5
665
AK168711.1
-
5
665
AK168750.1
-
5
668
AK168776.1
-
5
668
AK168887.1
-
5
667
AK168934.1
-
5
668
AK169093.1
-
5
665
AK169179.1
-
5
665
AK169236.1
-
5
665
AK169293.1
-
5
679
AK169329.1
-
5
668
AK169594.1
-
5
664
AK171434.1
-
5
666
AK178143.1
-
2
146
AK184919.1
-
2
146
AK190509.1
-
2
146
AK192182.1
-
2
147
AK192272.1
-
2
146
AK193850.1
-
2
146
AK193882.1
-
2
146
AK195240.1
-
2
146
AK198438.1
-
2
146
AK199142.1
-
2
146
AK199143.1
-
2
147
AK199178.1
-
2
146
AK199354.1
-
2
146
AK199410.1
-
2
179
AK199504.1
-
2
146
AK200094.1
-
2
146
AK200309.1
-
2
146
AK200579.1
-
2
146
AK201767.1
-
2
146
AK202859.1
-
2
146
AK203911.1
-
2
146
AK204890.1
-
2
146
AK205187.1
-
2
146
AK206134.1
-
2
146
AK206788.1
-
3
374
AK206953.1
-
2
146
AK209289.1
-
2
146
AK210012.1
-
3
212
AK211519.1
-
2
146
AK212081.1
-
3
210
AK212914.1
-
2
166
AK214003.1
-
3
193
AK216159.1
-
2
146
AK216732.1
-
2
146
AK216875.1
-
2
147
BC006722.1
-
5
689
3711399
BC066191.1
-
5
621
30467052
BC085486.1
-
5
653
30540914
BC089322.1
-
5
646
30538640
BC089457.1
-
5
624
30628505
BC094900.1
-
5
581
30063483
BC106169.1
-
3
345
BC106193.1
-
5
649
6329283
M19141.1
-
5
651
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_024351, XM_008764620, M11942, BC061547, FQ235115, and others.
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
protein folding
RNA splicing
protein refolding
negative regulation of transcription, DNA-templated
ATP metabolic process
positive regulation of mRNA splicing, via spliceosome
chaperone mediated protein folding requiring cofactor
regulation of cell cycle
chaperone-mediated protein folding
clathrin coat disassembly
negative regulation of fibril organization
nucleotide binding
G-protein coupled receptor binding
receptor binding
protein binding
ATP binding
ATPase activity
enzyme binding
MHC class II protein complex binding
heat shock protein binding
ubiquitin protein ligase binding
ATPase activity, coupled
ADP binding
poly(A) RNA binding
unfolded protein binding
C3HC4-type RING finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-sskjb:80/100.66.75.127:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0