Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tfec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031198.3 | NP_112475.1 | 6, 7 | 837 | ||
GenBank mRNA | AF077742.1 | - | 6, 7 | 837 | ||
AK137141.1 | - | 6, 7 | 802 | |||
AK156327.1 | - | 6, 7 | 750 | |||
AK159579.1 | - | 6, 7 | 770 | |||
BC052832.1 | - | 5, 6 | 725 | |||
BC064770.1 | - | 5, 6 | 713 | |||
BC098494.1 | - | 6, 7 | 722 | 6431607 |