Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290651.1 | NP_001277580.1 | 21 | 2530 | ||
NM_001290652.1 | NP_001277581.1 | 19 | 2350 | |||
GenBank mRNA | AF285577.1 | - | 18 | 1936 | ||
AK016533.1 | - | 18 | 1899 | |||
AK031490.1 | - | 21 | 2511 | |||
AK040521.1 | - | 19 | 2352 | |||
BC056382.1 | - | 21 | 2217 | 6853936 |