Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Qprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133686.1 | NP_598447.1 | 2 | 548 | ||
GenBank mRNA | AK010602.1 | - | 2 | 561 | ||
AK019119.1 | - | 2 | 563 | |||
BC011191.1 | - | 2 | 548 | 4196244 |