Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134183.1 | NP_598944.1 | 1 | 482 | ||
GenBank mRNA | BC120799.1 | - | 1 | 523 | 40129722 | |
BC120801.1 | - | 1 | 523 | 40129724 |