Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: REN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000537.3 | NP_000528.1 | 8, 9 | 1111 | ||
GenBank mRNA | BC033474.1 | - | 8, 9 | 1093 | 5188566 | |
BC047752.1 | - | 8, 9 | 1096 | 5590211 | ||
CR536498.1 | - | 8, 9 | 1067 | |||
X00063.1 | - | 6, 7 | 749 |