Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NLGN1 SIRNA Assays ›
          Gene Symbol:
          Nlgn1
          Gene Name:
          neuroligin 1
          Gene Aliases:
          6330415N05Rik, BB179718, NL1, Nlg1, mKIAA1070
          Chromosome Location:
          Chr.3: 25431841 - 26332056 on Build GRCm38
          UniGene ID:
          Mm.316080
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 88250

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nlgn1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001163387.1 NP_001156859.1 3 928
          NM_138666.3 NP_619607.2 5 1928
          GenBank mRNA AK122433.1 - 5 1929
          BC005523.1 - 2 599
          BC098461.1 - 3 928 30099449

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of respiratory gaseous exchange by neurological system process
          protein targeting
          cell adhesion
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          neuron cell-cell adhesion
          nervous system development
          synapse assembly
          positive regulation of circadian sleep/wake cycle, wakefulness
          synaptic vesicle targeting
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          neuron projection development
          positive regulation of synaptic transmission, GABAergic
          protein localization to synapse
          establishment of protein localization
          regulation of neuron differentiation
          synaptic vesicle transport
          rhythmic process
          cytoskeletal matrix organization at active zone
          modulation of synaptic transmission
          synapse organization
          protein homooligomerization
          protein heterotetramerization
          positive regulation of filopodium assembly
          positive regulation of synapse assembly
          positive regulation of synaptic transmission, glutamatergic
          inhibitory postsynaptic potential
          long-term synaptic potentiation
          positive regulation of dendritic spine development
          negative regulation of dendritic spine morphogenesis
          terminal button organization
          synaptic vesicle clustering
          postsynaptic membrane assembly
          presynaptic membrane assembly
          AMPA glutamate receptor clustering
          NMDA glutamate receptor clustering
          neurexin clustering involved in presynaptic membrane assembly
          postsynaptic density protein 95 clustering
          receptor localization to synapse
          positive regulation of ruffle assembly
          positive regulation of synaptic vesicle endocytosis
          positive regulation of protein localization to synapse
          positive regulation of intracellular signal transduction
          positive regulation of synaptic vesicle exocytosis
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
          positive regulation of excitatory postsynaptic potential
          positive regulation of synaptic vesicle clustering
          receptor activity
          protein binding
          PDZ domain binding
          neurexin family protein binding
          protein dimerization activity
          cell adhesion molecule binding
          carboxylic ester hydrolase activity
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-d79bl:80/100.66.79.68:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0