Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WT1 SIRNA Assays ›
Gene Symbol:
Wt1
Gene Name:
Wilms tumor 1 homolog
Gene Aliases:
D630046I19Rik, Wt-1
Chromosome Location:
Chr.2: 105126529 - 105173614 on Build GRCm38
UniGene ID:
Mm.389339
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
88493
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Wt1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_144783.2
NP_659032.3
1, 2
922
GenBank mRNA
M55512.1
-
1, 2
927
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_031534, XM_006234621, XM_006234620, XM_006234622, X69716.
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
vasculogenesis
metanephros development
ureteric bud development
branching involved in ureteric bud morphogenesis
kidney development
mesonephros development
regulation of organ formation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
apoptotic process
germ cell development
spermatogenesis
single fertilization
thorax and anterior abdomen determination
heart development
sex determination
negative regulation of cell proliferation
RNA splicing
gonad development
male gonad development
tissue development
regulation of gene expression
negative regulation of translation
negative regulation of cell growth
sperm motility
adrenal gland development
male genitalia development
epithelial cell differentiation
glomerulus development
glomerular basement membrane development
adrenal cortex formation
camera-type eye development
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
Sertoli cell development
mesenchymal to epithelial transition
positive regulation of heart growth
diaphragm development
cardiac muscle cell fate commitment
coronary vasculature development
visceral serous pericardium development
cellular response to cAMP
cellular response to gonadotropin stimulus
metanephric mesenchyme development
glomerular visceral epithelial cell differentiation
posterior mesonephric tubule development
metanephric S-shaped body morphogenesis
negative regulation of metanephric glomerular mesangial cell proliferation
seminiferous tubule development
negative regulation of mesenchymal cell apoptotic process involved in metanephros development
positive regulation of male gonad development
negative regulation of female gonad development
positive regulation of metanephric ureteric bud development
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
RNA binding
protein binding
zinc ion binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
C2H2 zinc finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0