Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ankrd33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144790.1 | NP_659039.1 | 3 | 378 | ||
GenBank mRNA | AK044543.1 | - | 4 | 563 | ||
AK044546.1 | - | 3 | 356 | |||
BC022726.1 | - | 3 | 378 | 4504539 | ||
FJ895380.1 | - | 4 | 539 |