Search
Search

Rpap2| Gene Symbol: Rpap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163461.2 | NP_001156933.1 | 2, 3 | 203 | ||
| NM_001163462.2 | NP_001156934.1 | 2, 3 | 260 | |||
| NM_001289569.1 | NP_001276498.1 | 2, 3 | 203 | |||
| NM_001289570.1 | NP_001276499.1 | 2, 3 | 203 | |||
| NM_144911.3 | NP_659160.2 | 2, 3 | 203 | |||
| GenBank mRNA | AK034418.1 | - | 2, 3 | 205 | ||
| AK049889.1 | - | 2, 3 | 260 | |||
| AK145830.1 | - | 2, 3 | 178 | |||
| AK161901.1 | - | 2, 3 | 184 | |||
| AK166574.1 | - | 2, 3 | 126 | |||
| BC021895.1 | - | 2, 3 | 164 | 3481916 | ||