Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tapbpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145391.2 | NP_663366.2 | 2 | 481 | ||
GenBank mRNA | BC017613.1 | - | 2 | 481 | 4912570 | |
BX517366.1 | - | 2 | 334 |