Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap1gds1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040690.2 | NP_001035780.1 | 2 | 243 | ||
NM_001286759.1 | NP_001273688.1 | 2 | 243 | |||
NM_145544.3 | NP_663519.2 | 2 | 243 | |||
GenBank mRNA | AK052125.1 | - | 2 | 191 | ||
AK154852.1 | - | 2 | 182 | |||
AK155793.1 | - | 2 | 182 | |||
AK158465.1 | - | 2 | 236 | |||
AK160989.1 | - | 2 | 179 | |||
AK164159.1 | - | 2 | 155 | |||
AK166315.1 | - | 2 | 176 | |||
AK171900.1 | - | 2 | 182 | |||
AK172052.1 | - | 2 | 245 | |||
BC011279.1 | - | 2 | 112 | 4008751 | ||
BY015015.1 | - | 2 | 242 |