Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4k2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145501.2 | NP_663476.1 | 7 | 1209 | ||
GenBank mRNA | AK030725.1 | - | 7 | 1174 | ||
AK165136.1 | - | 1 | 1265 | |||
AK185438.1 | - | 3 | 230 | |||
BC089309.1 | - | 6 | 1898 | |||
BC110363.1 | - | 7 | 1171 | 5346917 |