Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rap1gds1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040690.2 | NP_001035780.1 | 9, 10 | 1204 | ||
NM_001286759.1 | NP_001273688.1 | 9, 10 | 1204 | |||
NM_145544.3 | NP_663519.2 | 8, 9 | 1057 | |||
GenBank mRNA | AK052125.1 | - | 8, 9 | 1005 | ||
AK154852.1 | - | 9, 10 | 1190 | |||
AK155793.1 | - | 9, 10 | 1190 | |||
AK160989.1 | - | 9, 10 | 1140 | |||
AK164159.1 | - | 9, 10 | 1116 | |||
AK166315.1 | - | 9, 10 | 1137 | |||
AK172052.1 | - | 9, 10 | 1206 | |||
BC011279.1 | - | 8, 9 | 926 | 4008751 | ||
CN702379.1 | - | 17 |