Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Polr2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145632.2 | NP_663607.1 | 3, 4 | 355 | ||
GenBank mRNA | AK146429.1 | - | 3, 4 | 380 | ||
AK161617.1 | - | 3, 4 | 376 | |||
BC002306.1 | - | 3, 4 | 355 | 3591968 | ||
BC092087.1 | - | 3, 4 | 265 | 5042770 | ||
BC125612.1 | - | 3, 4 | 339 | 40130127 | ||
BC125638.1 | - | 3, 4 | 339 | 40130153 |