Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CRH SIRNA Assays ›
          Gene Symbol:
          CRH
          Gene Name:
          corticotropin releasing hormone
          Gene Aliases:
          CRF, CRH1
          Chromosome Location:
          Chr.8: 66176377 - 66186685 on Build GRCh38
          UniGene ID:
          Hs.75294
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 9004

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CRH
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000756.3 NP_000747.1 2 1200
          XM_017013090.1 XP_016868579.1 6 1538
          XM_017013091.1 XP_016868580.1 6 1601
          XM_017013092.1 XP_016868581.1 6 1539
          XM_017013093.1 XP_016868582.1 6 1678
          GenBank mRNA AK075431.1 - 2 1146
          BC011031.1 - 2 1022 4280754

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          synaptic transmission, dopaminergic
          glucocorticoid biosynthetic process
          inflammatory response
          signal transduction
          chemical synaptic transmission
          female pregnancy
          parturition
          learning or memory
          positive regulation of cell proliferation
          associative learning
          hormone-mediated apoptotic signaling pathway
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of norepinephrine secretion
          positive regulation of circadian sleep/wake cycle, wakefulness
          positive regulation of cell death
          regulation of serotonin secretion
          diterpenoid metabolic process
          hypothalamus development
          lung development
          adrenal gland development
          positive regulation of cAMP biosynthetic process
          negative regulation of epinephrine secretion
          negative regulation of luteinizing hormone secretion
          locomotory exploration behavior
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          response to immobilization stress
          negative regulation of circadian sleep/wake cycle, REM sleep
          response to drug
          response to estrogen
          response to ethanol
          response to ether
          negative regulation of blood pressure
          response to pain
          ion homeostasis
          response to corticosterone
          positive regulation of corticotropin secretion
          positive regulation of cortisol secretion
          long-term synaptic potentiation
          positive regulation of digestive system process
          negative regulation of cell death
          negative regulation of glucagon secretion
          cellular response to cocaine
          cellular response to dexamethasone stimulus
          positive regulation of calcium ion import
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          positive regulation of corticosterone secretion
          positive regulation of behavioral fear response
          receptor binding
          hormone activity
          neuropeptide hormone activity
          protein binding
          corticotropin-releasing hormone activity
          corticotropin-releasing hormone receptor 1 binding
          corticotropin-releasing hormone receptor 2 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0