Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rfwd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146218.4 | NP_666330.2 | 2 | 131 | ||
GenBank mRNA | AK137499.1 | - | 2 | 111 | ||
AK137698.1 | - | 2 | 94 | |||
AK139032.1 | - | 2 | 111 | |||
AK146451.1 | - | 2 | 101 | |||
BC023694.1 | - | 2 | 136 | 5344528 | ||
BC096602.1 | - | 2 | 94 | 4954068 |