Search
Search

Mapre2| Gene Symbol: Mapre2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162941.1 | NP_001156413.1 | 8 | 1809 | ||
| NM_001162942.1 | NP_001156414.1 | 8 | 1752 | |||
| NM_153058.4 | NP_694698.3 | 7 | 1860 | |||
| GenBank mRNA | AK028823.1 | - | 8 | 1810 | ||
| AK038562.1 | - | 7 | 1847 | |||
| AK045493.1 | - | 8 | 1708 | |||
| AK045904.1 | - | 7 | 1835 | |||
| AK046095.1 | - | 7 | 1803 | |||
| AK046221.1 | - | 7 | 1827 | |||
| AK148954.1 | - | 7 | 1835 | |||
| BC025804.1 | - | 7 | 1807 | 5101879 | ||
| BC027056.1 | - | 7 | 1826 | 5361238 | ||
| BC035254.1 | - | 7 | 1812 | 5363176 | ||