Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Ntmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC027220.1 | - | 1214 | 3991341 |