Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172283.3 | NP_758487.2 | 10 | 972 | ||
GenBank mRNA | AJ297482.4 | - | 10 | 943 | ||
AJ534942.1 | - | 10 | 943 | |||
AK131165.1 | - | 2 | 652 | |||
AK161763.1 | - | 10 | 974 | |||
AK164340.1 | - | 10 | 941 | |||
AY223865.1 | - | 10 | 943 | |||
BC037698.1 | - | 9 | 921 | 4190449 |