Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172399.3 | NP_765987.2 | 2 | 581 | ||
GenBank mRNA | AK029210.1 | - | 2 | 582 | ||
BC047154.1 | - | 2 | 555 | 6489681 |