Search Thermo Fisher Scientific
Gene Symbol: Itih5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172471.2 | NP_766059.1 | 4 | 508 | ||
GenBank mRNA | AK077381.1 | - | 431 | |||
AK087428.1 | - | 4 | 436 | |||
AK134707.1 | - | 4 | 436 | |||
AK140756.1 | - | 4 | 509 | |||
BC043314.1 | - | 4 | 451 | 5150581 | ||
BC062196.1 | - | 4 | 492 | 3490706 |