Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Nipal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172524.3 | NP_766112.1 | 2 | 249 | ||
GenBank mRNA | AK035561.1 | - | 2 | 250 | ||
AK035736.1 | - | 2 | 226 | |||
BC139819.1 | - | 2 | 240 | 40141293 |