Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rsbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172684.2 | NP_766272.2 | 2 | 1246 | ||
GenBank mRNA | AB101658.1 | - | 2 | 1248 | ||
AK087685.1 | - | 1 | 212 | |||
AK122577.1 | - | 2 | 781 | |||
AK134093.1 | - | 2 | 246 | |||
AK138603.1 | - | 2 | 790 | |||
BC094667.1 | - | 2 | 771 | |||
BC139213.1 | - | 2 | 1215 | 8862235 | ||
BC139215.1 | - | 2 | 1215 | 8862237 |