Search
Search

Galnt12| Gene Symbol: Galnt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172693.3 | NP_766281.1 | 2 | 520 | ||
| GenBank mRNA | AB175680.1 | - | 2 | 500 | ||
| AK033638.1 | - | 2 | 521 | |||
| AK042133.1 | - | 2 | 553 | |||
| AK082014.1 | - | 2 | 566 | |||
| AK156751.1 | - | 2 | 550 | |||
| BC025639.1 | - | 1 | 242 | |||
| BC056425.1 | - | 2 | 531 | 5716909 | ||