Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dip2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159361.1 | NP_001152833.1 | 7 | 1025 | ||
NM_172819.3 | NP_766407.2 | 2 | 240 | |||
GenBank mRNA | AK030064.1 | - | 2 | 241 | ||
AK147562.1 | - | 7 | 961 | |||
BC137857.1 | - | 2 | 227 | 8860876 | ||
BC137858.1 | - | 2 | 227 | 8860877 |