Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Muc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290786.1 | NP_001277715.1 | 2 | 592 | ||
NM_172979.3 | NP_766567.1 | 2 | 592 | |||
GenBank mRNA | AK052840.1 | - | 2 | 479 | ||
AK142590.1 | - | 2 | 479 | |||
AY198336.1 | - | 2 | 508 | |||
AY198337.1 | - | 2 | 475 | |||
BC094506.1 | - | 2 | 475 | 5341608 | ||
BC116796.1 | - | 2 | 471 | 40126115 | ||
BC119176.1 | - | 2 | 471 | 8733925 |