Search
Search

Galnt13| Gene Symbol: Galnt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173030.2 | NP_766618.2 | 3, 4 | 660 | ||
| GenBank mRNA | AB082928.1 | - | 1, 2 | 148 | ||
| AK030726.1 | - | 2, 3 | 590 | |||
| AK038387.1 | - | 3, 4 | 661 | |||
| AK045417.1 | - | 3, 4 | 661 | |||
| BC131651.1 | - | 3, 4 | 339 | 40110517 | ||
| BC131652.1 | - | 2, 3 | 370 | 40110518 | ||