Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGER SIRNA Assays ›
          Gene Symbol:
          AGER
          Gene Name:
          advanced glycosylation end product-specific receptor
          Gene Aliases:
          RAGE, SCARJ1
          Chromosome Location:
          Chr.6: 32180968 - 32185247 on Build GRCh38
          UniGene ID:
          Hs.534342
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 9534

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AGER
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136.4 NP_001127.1 3 419
          NM_001206929.1 NP_001193858.1 3 419
          NM_001206932.1 NP_001193861.1 3 377
          NM_001206934.1 NP_001193863.1 3 419
          NM_001206936.1 NP_001193865.1 3 419
          NM_001206940.1 NP_001193869.1 3 419
          NM_001206954.1 NP_001193883.1 3 419
          NM_001206966.1 NP_001193895.1 3 419
          NM_172197.2 NP_751947.1 3 377
          NR_038190.1 2 602
          XM_017010328.1 XP_016865817.1 3 1344
          GenBank mRNA AB036432.1 - 3 343
          AB061668.1 - 3 343
          AB061669.1 - 2 526
          AJ133822.1 - 3 292
          AK298100.1 - 3 421
          AK313178.1 - 3 347
          AY755619.1 - 3 329
          AY755620.1 - 3 329
          AY755621.1 - 3 329
          AY755622.1 - 3 329
          AY755623.1 - 3 329
          AY755624.1 - 3 287
          AY755625.1 - 3 329
          AY755628.1 - 3 329
          BC020669.1 - 3 338 4718076
          DQ104251.1 - 3 329
          DQ104252.1 - 3 287
          DQ104253.1 - 3 329
          DQ104254.1 - 2 512
          EU117141.1 - 3 329
          EU428812.1 - 2 502
          EU428813.1 - 2 502
          EU428815.1 - 2 502
          EU428816.1 - 2 502
          EU826616.1 - 3 319
          EU826617.1 - 3 319
          EU826618.1 - 3 319
          EU826619.1 - 3 319
          EU826620.1 - 3 319
          FJ545655.1 - 3 339
          GU139370.1 - 1 654
          GU139371.1 - 1 654
          GU139372.1 - 2 524
          GU139373.1 - 1 655
          GU139374.1 - 1 653
          GU139375.1 - 2 524
          GU139376.1 - 2 524
          GU139377.1 - 2 525
          GU139378.1 - 3 341
          HQ148133.1 - 3 343
          HQ148134.1 - 3 343
          HQ148135.1 - 2 523
          KC692917.1 - 3 319
          M91211.1 - 3 319

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cell adhesion molecule immunoglobulin superfamily cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of T cell mediated cytotoxicity
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          negative regulation of endothelial cell proliferation
          inflammatory response
          negative regulation of cell adhesion
          cell surface receptor signaling pathway
          JAK-STAT cascade
          brain development
          glycoprotein metabolic process
          response to wounding
          response to fructose
          positive regulation of autophagy
          negative regulation of endothelial cell migration
          positive regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          positive regulation of fibroblast migration
          response to activity
          positive regulation of smooth muscle cell migration
          lung development
          neuron projection development
          negative regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          negative regulation of collagen biosynthetic process
          response to vitamin A
          response to genistein
          negative regulation of osteoblast proliferation
          cellular response to drug
          positive regulation of activated T cell proliferation
          positive regulation of neuron apoptotic process
          innate immune response
          positive regulation of fibroblast proliferation
          positive regulation of smooth muscle cell proliferation
          positive regulation of inflammatory response
          positive regulation of NF-kappaB transcription factor activity
          response to methylglyoxal
          calcium ion homeostasis
          response to hyperoxia
          positive regulation of phagocytosis, engulfment
          transdifferentiation
          cellular response to hydrogen peroxide
          cellular response to glucose stimulus
          cellular response to fatty acid
          cellular response to organic cyclic compound
          response to selenite ion
          positive regulation of potassium ion transmembrane transporter activity
          positive regulation of endothelial cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          regulation of CD4-positive, alpha-beta T cell activation
          positive regulation of type B pancreatic cell apoptotic process
          positive regulation of dendritic cell differentiation
          receptor activity
          transmembrane signaling receptor activity
          protein binding
          identical protein binding
          S100 protein binding
          advanced glycation end-product receptor activity
          high mobility group box 1 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0