Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Peak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172924.3 | NP_766512.2 | 4 | 824 | ||
GenBank mRNA | AK035440.1 | - | 4 | 814 | ||
AK048912.1 | - | 4 | 813 | |||
AK090133.1 | - | 4 | 840 |