Search
Search

Map1s| Gene Symbol: Map1s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173013.3 | NP_766601.2 | 5 | 1286 | ||
| GenBank mRNA | AK032300.1 | - | 5 | 1274 | ||
| AK042159.1 | - | 5 | 1283 | |||
| AK082854.1 | - | 5 | 1281 | |||
| AK158860.1 | - | 3 | 1055 | |||
| BC052828.1 | - | 1 | 133 | |||
| BC058166.1 | - | 5 | 1267 | |||
| BC079598.1 | - | 3 | 2647 | |||
| BC150729.1 | - | 5 | 1328 | 9087640 | ||
| DQ387862.1 | - | 5 | 1274 | |||