Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 4933427D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175017.3 | NP_778182.1 | 4 | 286 | ||
GenBank mRNA | AK030235.1 | - | 4 | 224 | ||
BC098480.1 | - | 4 | 286 | 30048571 |