Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDH13 SIRNA Assays ›
          Gene Symbol:
          CDH13
          Gene Name:
          cadherin 13
          Gene Aliases:
          CDHH, P105
          Chromosome Location:
          Chr.16: 82626794 - 83796610 on Build GRCh38
          UniGene ID:
          Hs.654386
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 9590

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDH13
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001220488.1 NP_001207417.1 4 590
          NM_001220489.1 NP_001207418.1 3 484
          NM_001220491.1 NP_001207420.1 3 484
          NM_001220492.1 NP_001207421.1 3 484
          NM_001257.4 NP_001248.1 3 484
          XM_017022848.1 XP_016878337.1 4 507
          XM_017022849.1 XP_016878338.1 4 552
          GenBank mRNA AK298612.1 - 3 310
          AK304733.1 - 4 345
          AU123178.1 - 3 309
          BC028624.2 - 3 276 4825734
          BC030653.2 - 3 276 4811561
          BI756586.1 - 3 287
          CV024366.1 - 3 185
          DC376661.1 - 3 484
          EU190357.1 - 3 311
          EU190358.1 - 3 311
          L34058.1 - 3 629
          U59288.1 - 3 401

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          positive regulation of endothelial cell proliferation
          positive regulation of cell-matrix adhesion
          sprouting angiogenesis
          homophilic cell adhesion via plasma membrane adhesion molecules
          negative regulation of cell adhesion
          Rho protein signal transduction
          negative regulation of cell proliferation
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          Rac protein signal transduction
          lamellipodium assembly
          regulation of endocytosis
          positive regulation of cell migration
          adherens junction organization
          regulation of epidermal growth factor receptor signaling pathway
          endothelial cell migration
          keratinocyte proliferation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of smooth muscle cell proliferation
          positive regulation of calcium-mediated signaling
          positive regulation of positive chemotaxis
          localization within membrane
          low-density lipoprotein particle mediated signaling
          calcium ion binding
          low-density lipoprotein particle binding
          protein homodimerization activity
          cadherin binding
          adiponectin binding
          lipoprotein particle binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0