Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Insc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173767.3 | NP_776128.2 | 8 | 1264 | ||
GenBank mRNA | AK014452.1 | - | 7 | 1025 | ||
AK050143.1 | - | 5 | 759 | |||
AK204816.1 | - | 2 | 218 | |||
AK211253.1 | - | 2 | 212 | |||
BC026405.1 | - | 8 | 1151 | 4211657 | ||
BC089586.1 | - | 5 | 768 | |||
BC139196.1 | - | 5 | 652 | 8862218 | ||
BC139197.1 | - | 5 | 652 | 8862219 | ||
DQ205645.1 | - | 7 | 977 |