Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rnf139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175226.4 | NP_780435.1 | 2 | 771 | ||
GenBank mRNA | AK028400.1 | - | 2 | 724 | ||
AK029069.1 | - | 2 | 774 | |||
AK033506.1 | - | 2 | 705 | |||
BC052901.1 | - | 2 | 720 | 30070643 |