Search Thermo Fisher Scientific
Gene Symbol: Snx33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175483.5 | NP_780692.2 | 1 | 1665 | ||
GenBank mRNA | AK048566.1 | - | 1 | 170 | ||
AK053761.1 | - | 1 | 200 | |||
AK142274.1 | - | 1 | 1666 | |||
AK179703.1 | - | 1 | 332 | |||
AK179871.1 | - | 1 | 329 | |||
AK191380.1 | - | 1 | 371 | |||
BC096472.1 | - | 1 | 415 | 3501525 |