Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Snrnp200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177214.4 | NP_796188.2 | 4, 5 | 656 | ||
GenBank mRNA | AK039567.1 | - | 4, 5 | 662 | ||
AK081920.1 | - | 4, 5 | 675 | |||
BC063261.1 | - | 4, 5 | 656 | 6408607 |