Search
Search

Depdc5| Gene Symbol: Depdc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025426.2 | NP_001020597.1 | 6, 7 | 756 | ||
| NM_001170567.1 | NP_001164038.1 | 5, 6 | 453 | |||
| NM_177786.4 | NP_808454.2 | 5, 6 | 453 | |||
| GenBank mRNA | AK043137.1 | - | 5, 6 | 405 | ||
| AK052458.1 | - | 5, 6 | 441 | |||
| AK078771.1 | - | 5, 6 | 398 | |||
| AK129180.1 | - | 5, 6 | 417 | |||
| AK136321.1 | - | 5, 6 | 398 | |||
| AK166366.1 | - | 6, 7 | 718 | |||
| AK172587.1 | - | 5, 6 | 405 | |||
| BC072604.1 | - | 6, 7 | 756 | 6831032 | ||