Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177818.3 | NP_808486.2 | 4 | 981 | ||
GenBank mRNA | AK048392.1 | - | 4 | 876 | ||
EF494899.1 | - | 819 | ||||
EF494900.1 | - | 819 | ||||
EF494901.1 | - | 664 | ||||
EF494902.1 | - | 664 |