Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACTN2 SIRNA Assays ›
          Gene Symbol:
          ACTN2
          Gene Name:
          actinin alpha 2
          Gene Aliases:
          CMD1AA, CMH23
          Chromosome Location:
          Chr.1: 236686454 - 236764631 on Build GRCh38
          UniGene ID:
          Hs.498178
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS100132
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACTN2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001103.3 NP_001094.1 10, 11 1319
          NM_001278343.1 NP_001265272.1 10, 11 1319
          NM_001278344.1 NP_001265273.1 12, 13 1516
          GenBank mRNA AB209521.1 - 12, 13 1460
          AK294906.1 - 8, 9 830
          AK297469.1 - 4, 5 464
          AK297618.1 - 3, 4 401
          AK315250.1 - 10, 11 1319
          BC047901.2 - 10, 11 1251 6199155
          BC051770.1 - 10, 11 1264 6198688
          M86406.1 - 10, 11 1272

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          platelet degranulation
          cell adhesion
          microspike assembly
          muscle filament sliding
          regulation of membrane potential
          regulation of apoptotic process
          negative regulation of potassium ion transport
          positive regulation of potassium ion transport
          positive regulation of GTPase activity
          sarcomere organization
          focal adhesion assembly
          protein homotetramerization
          actin filament uncapping
          cardiac muscle cell development
          phospholipase C-activating angiotensin-activated signaling pathway
          establishment of protein localization to plasma membrane
          negative regulation of potassium ion transmembrane transporter activity
          positive regulation of potassium ion transmembrane transporter activity
          regulation of nucleic acid-templated transcription
          negative regulation of protein localization to cell surface
          positive regulation of receptor activity
          positive regulation of endocytic recycling
          positive regulation of cation channel activity
          Ras guanyl-nucleotide exchange factor activity
          integrin binding
          calcium ion binding
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          cytoskeletal protein binding
          structural constituent of muscle
          protein domain specific binding
          LIM domain binding
          thyroid hormone receptor coactivator activity
          titin binding
          identical protein binding
          protein homodimerization activity
          ion channel binding
          protein dimerization activity
          actin filament binding
          FATZ binding
          titin Z domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0