Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADSL SIRNA Assays ›
          Gene Symbol:
          ADSL
          Gene Name:
          adenylosuccinate lyase
          Gene Aliases:
          AMPS, ASASE, ASL
          Chromosome Location:
          Chr.22: 40346500 - 40387408 on Build GRCh38
          UniGene ID:
          Hs.75527
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS100271
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADSL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000026.3 NP_000017.1 5, 6 716
          NM_001123378.2 NP_001116850.1 5, 6 716
          NM_001317923.1 NP_001304852.1 5, 6 661
          NR_134256.1 5, 6 716
          XM_011529977.2 XP_011528279.1 5, 6 716
          XM_011529979.1 XP_011528281.1 5, 6 716
          XM_011529980.2 XP_011528282.1 5, 6 716
          XM_017028636.1 XP_016884125.1 4, 5 653
          XM_017028637.1 XP_016884126.1 4, 5 636
          XM_017028638.1 XP_016884127.1 3, 4 573
          XM_017028639.1 XP_016884128.1 3, 4 573
          XM_017028640.1 XP_016884129.1 1, 2 71
          GenBank mRNA AF067853.1 - 5, 6 712
          AF067854.1 - 5, 6 712
          AK225042.1 - 5, 6 684
          AK291091.1 - 5, 6 687
          AK298187.1 - 5, 6 730
          AK300705.1 - 3, 4 493
          AK308662.1 - 1, 2 71
          BC000253.1 - 5, 6 686 3352079
          BC018881.2 - 5, 6 686
          CR456368.1 - 5, 6 690
          KJ534763.1 - 5, 6 657
          KJ535007.1 - 5, 6 657
          KJ535020.1 - 5, 6 657
          KJ535041.1 - 2, 3 328
          KJ535056.1 - 5, 6 602
          S60710.1 - 5, 6 659
          X65867.1 - 5, 6 659

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          lyase

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleotide biosynthetic process
          AMP biosynthetic process
          'de novo' IMP biosynthetic process
          purine ribonucleoside monophosphate biosynthetic process
          'de novo' AMP biosynthetic process
          protein tetramerization
          N6-(1,2-dicarboxyethyl)AMP AMP-lyase (fumarate-forming) activity
          (S)-2-(5-amino-1-(5-phospho-D-ribosyl)imidazole-4-carboxamido)succinate AMP-lyase (fumarate-forming) activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-79lxv:80/100.66.77.52:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0