Search
Search

ANPEP| Gene Symbol: ANPEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001150.2 | NP_001141.2 | 19, 20 | 2970 | ||
| XM_005254892.4 | XP_005254949.1 | 19, 20 | 2978 | |||
| XM_011521473.1 | XP_011519775.1 | 19, 20 | 3121 | |||
| GenBank mRNA | AB209918.1 | - | 19, 20 | 2966 | ||
| AK298134.1 | - | 13, 14 | 1440 | |||
| AK298242.1 | - | 13, 14 | 1491 | |||
| AK298339.1 | - | 14, 15 | 1551 | |||
| AK300946.1 | - | 15, 16 | 1701 | |||
| BC058928.1 | - | 18, 19 | 2771 | 6020902 | ||
| M22324.1 | - | 18, 19 | 2798 | |||
| X13276.1 | - | 18, 19 | 2702 | |||