Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANXA1 SIRNA Assays ›
          Gene Symbol:
          ANXA1
          Gene Name:
          annexin A1
          Gene Aliases:
          ANX1, LPC1
          Chromosome Location:
          Chr.9: 73151731 - 73170393 on Build GRCh38
          UniGene ID:
          Hs.494173
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS100502
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANXA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000700.2 NP_000691.1 3 366
          XM_011518609.1 XP_011516911.1 3 293
          XM_017014657.1 XP_016870146.1 3 312
          GenBank mRNA AB451274.1 - 2 158
          AB451401.1 - 2 158
          AK074480.1 - 2 1040
          AK296808.1 - 3 232
          BC001275.1 - 3 210 3459615
          BC035993.1 - 3 232 4662939
          BT019896.1 - 2 158
          BT019916.1 - 2 158
          BT019917.1 - 2 158
          CR407684.1 - 2 158
          CX870105.1 - 3 366
          X05908.1 - 3 232

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          calcium-binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA strand renaturation
          neutrophil homeostasis
          adaptive immune response
          monocyte chemotaxis
          regulation of leukocyte migration
          phagocytosis
          inflammatory response
          signal transduction
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          regulation of cell shape
          response to X-ray
          myoblast migration involved in skeletal muscle regeneration
          peptide cross-linking
          insulin secretion
          keratinocyte differentiation
          prostate gland development
          endocrine pancreas development
          positive regulation of vesicle fusion
          positive regulation of prostaglandin biosynthetic process
          actin cytoskeleton reorganization
          response to estradiol
          DNA duplex unwinding
          regulation of interleukin-1 production
          positive regulation of interleukin-2 production
          positive regulation of neutrophil apoptotic process
          DNA rewinding
          gliogenesis
          positive regulation of T cell proliferation
          response to drug
          negative regulation of apoptotic process
          response to peptide hormone
          estrous cycle
          innate immune response
          positive regulation of T-helper 1 cell differentiation
          negative regulation of T-helper 2 cell differentiation
          negative regulation of exocytosis
          alpha-beta T cell differentiation
          regulation of hormone secretion
          arachidonic acid secretion
          regulation of inflammatory response
          cellular response to hydrogen peroxide
          hepatocyte differentiation
          prolactin secretion
          response to interleukin-1
          cellular response to glucocorticoid stimulus
          granulocyte chemotaxis
          positive regulation of wound healing
          neutrophil clearance
          positive regulation of G1/S transition of mitotic cell cycle
          negative regulation of phospholipase A2 activity
          negative regulation of interleukin-8 secretion
          single-stranded DNA binding
          single-stranded RNA binding
          helicase activity
          receptor binding
          structural molecule activity
          calcium ion binding
          protein binding
          phospholipid binding
          calcium-dependent phospholipid binding
          phospholipase A2 inhibitor activity
          protein binding, bridging
          double-stranded DNA-dependent ATPase activity
          annealing helicase activity
          protein homodimerization activity
          calcium-dependent protein binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0