Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP6V0A1 SIRNA Assays ›
Gene Symbol:
ATP6V0A1
Gene Name:
ATPase H+ transporting V0 subunit a1
Gene Aliases:
ATP6N1, ATP6N1A, Stv1, VPP1, Vph1, a1
Chromosome Location:
Chr.17: 42458844 - 42522579 on Build GRCh38
UniGene ID:
Hs.463074
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS100862
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATP6V0A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130020.1
NP_001123492.1
12, 13
1494
NM_001130021.1
NP_001123493.1
12, 13
1473
NM_005177.3
NP_005168.2
12, 13
1473
XM_005257459.1
XP_005257516.1
12, 13
1476
XM_005257461.1
XP_005257518.1
12, 13
1476
XM_005257463.2
XP_005257520.1
12, 13
1476
XM_011524908.1
XP_011523210.1
12, 13
1476
XM_017024766.1
XP_016880255.1
12, 13
1476
XM_017024767.1
XP_016880256.1
12, 13
1439
XM_017024768.1
XP_016880257.1
12, 13
1439
XM_017024769.1
XP_016880258.1
12, 13
1439
XM_017024770.1
XP_016880259.1
12, 13
1439
XM_017024771.1
XP_016880260.1
12, 13
1439
XM_017024772.1
XP_016880261.1
11, 12
1286
XM_017024773.1
XP_016880262.1
11, 12
1867
XM_017024774.1
XP_016880263.1
12, 13
1467
GenBank mRNA
AK223554.1
-
12, 13
1440
AK294512.1
-
11, 12
1300
AK294789.1
-
8, 9
954
AK295682.1
-
12, 13
1397
AK299784.1
-
11, 12
1312
AK316305.1
-
12, 13
1464
BC032398.1
-
12, 13
1405
5195776
BX648978.1
-
12, 13
1456
CR627443.1
-
5, 6
381
L78933.1
-
12, 13
1475
Z71460.1
-
12, 13
1475
Back To Top
More Information
Panther Classification:
Molecular Function -
ATP synthase
Gene Ontology Categories:
Function(s)
Process(es)
vacuolar acidification
insulin receptor signaling pathway
ATP synthesis coupled proton transport
ATP hydrolysis coupled proton transport
regulation of macroautophagy
transferrin transport
ion transmembrane transport
vacuolar proton-transporting V-type ATPase complex assembly
phagosome acidification
toxin transport
protein binding
proton-transporting ATPase activity, rotational mechanism
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-srbv6:80/100.66.75.104:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0