Search
Search

C2| Gene Symbol: C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000063.5 | NP_000054.2 | 15 | 2151 | ||
| NM_001145903.2 | NP_001139375.1 | 13 | 1755 | |||
| NM_001178063.2 | NP_001171534.1 | 11 | 1301 | |||
| NM_001282457.1 | NP_001269386.1 | 11 | 1358 | |||
| NM_001282458.1 | NP_001269387.1 | 15 | 2179 | |||
| GenBank mRNA | AK096258.1 | - | 12 | 1456 | ||
| AK222537.1 | - | 15 | 1903 | |||
| AK222783.1 | - | 15 | 1875 | |||
| AK298311.1 | - | 11 | 1301 | |||
| AK298808.1 | - | 11 | 1358 | |||
| AK300892.1 | - | 13 | 1515 | |||
| AK300930.1 | - | 15 | 1917 | |||
| AK312581.1 | - | 15 | 1985 | |||
| BC043484.1 | - | 15 | 1886 | 5763774 | ||
| BX537504.1 | - | 9 | 1888 | |||
| X04481.1 | - | 15 | 1911 | |||