Search
Search

CACNA1A| Gene Symbol: CACNA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000068.3 | NP_000059.3 | 24, 25 | 4241 | ||
| NM_001127221.1 | NP_001120693.1 | 24, 25 | 4232 | |||
| NM_001127222.1 | NP_001120694.1 | 24, 25 | 4229 | |||
| NM_001174080.1 | NP_001167551.1 | 24, 25 | 4232 | |||
| NM_023035.2 | NP_075461.2 | 24, 25 | 4241 | |||
| GenBank mRNA | AB035726.1 | - | 24, 25 | 4031 | ||
| AB035727.2 | - | 24, 25 | 3993 | |||
| AF004883.1 | - | 24, 25 | 4241 | |||
| AF004884.1 | - | 24, 25 | 4241 | |||
| FJ040507.1 | - | 24, 25 | 3996 | |||
| U79663.1 | - | 1, 2 | 62 | |||
| U79664.1 | - | 1, 2 | 62 | |||
| U79665.1 | - | 1, 2 | 62 | |||
| U79666.1 | - | 24, 25 | 4213 | |||
| U79667.1 | - | 1, 2 | 62 | |||
| U79668.1 | - | 1, 2 | 62 | |||
| X99897.1 | - | 24, 25 | 4274 | |||