Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KYAT1 SIRNA Assays ›
Gene Symbol:
KYAT1
Gene Name:
kynurenine aminotransferase 1
Gene Aliases:
CCBL1, GTK, KAT1, KATI
Chromosome Location:
Chr.9: 128832942 - 128882551 on Build GRCh38
UniGene ID:
Hs.495250
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS101435
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KYAT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122671.1
NP_001116143.1
6
692
NM_001122672.1
NP_001116144.1
5
548
NM_001287390.1
NP_001274319.1
8
1089
NM_004059.4
NP_004050.3
6
698
NR_109829.1
6
698
XM_006717311.3
XP_006717374.1
7
884
XM_011519165.2
XP_011517467.1
8
1060
XM_011519167.2
XP_011517469.2
6
664
XM_011519168.1
XP_011517470.1
5
523
XM_011519169.1
XP_011517471.1
6
792
XM_011519170.2
XP_011517472.1
6
1160
XM_017015260.1
XP_016870749.1
7
844
XM_017015261.1
XP_016870750.1
8
1066
XM_017015262.1
XP_016870751.1
8
1555
XM_017015263.1
XP_016870752.1
8
970
XM_017015264.1
XP_016870753.1
7
877
XM_017015265.1
XP_016870754.1
6
1254
XM_017015266.1
XP_016870755.1
7
885
XM_017015267.1
XP_016870756.1
6
706
XM_017015268.1
XP_016870757.1
7
868
GenBank mRNA
AK094505.1
-
6
1241
AK291178.1
-
6
686
AK297995.1
-
8
945
AK314427.1
-
6
568
AU123740.1
-
6
676
BC021262.2
-
6
659
4137942
BC033685.1
-
5
402
5193675
BI755163.1
-
5
402
EL733697.1
-
4
363
X82224.1
-
5
513
Back To Top
More Information
Panther Classification:
Molecular Function -
transaminase
Gene Ontology Categories:
Function(s)
Process(es)
L-phenylalanine catabolic process
tryptophan catabolic process
cellular modified amino acid metabolic process
cellular amino acid biosynthetic process
kynurenine metabolic process
L-kynurenine catabolic process
protein binding
transaminase activity
kynurenine-oxoglutarate transaminase activity
pyridoxal phosphate binding
protein homodimerization activity
L-phenylalanine:pyruvate aminotransferase activity
glutamine-phenylpyruvate transaminase activity
cysteine-S-conjugate beta-lyase activity
L-glutamine:pyruvate aminotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0